missing translation for 'onlineSavingsMsg'
Learn More

Contactin 4, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen OSC00014W

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11590443

  • £257.00 / 100ĀµL


Estimated Shipment
15-06-2018
Add to basket

Description

Description

Reconstitute with 100 ul of distilled water.
Specifications

Specifications

Contactin 4
Polyclonal
whole serum with no preservative
Q8IWV2
Brain-derived immunoglobulin superfamily protein 2, AXCAM, contactin 4, BIG-2, SCA16, CNTN4A, MGC33615
Rabbit
100μL
Antibody
Polyclonal
Human
ImmunohistochemistryĆ¾Western Blot
Unconjugated
CNTN4
Serum
CNTN4
A synthetic peptide from human Contactin-4
store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
152330
Videos