missing translation for 'onlineSavingsMsg'
Learn More

Contactin 2, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen OSC00022W

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11570453

  • £257.00 / 100ĀµL


Estimated Shipment
15-06-2018
Add to basket

Description

Description

Reconstitute with 100 ul of distilled water.
Specifications

Specifications

Contactin 2
Polyclonal
whole serum with no preservative
Q02246
Axonin-1, Axonal glycoprotein TAG-1, Transient axonal glycoprotein 1, TAX-1, CNTN2, AXT, TAG1, TAX1, MGC157722, DKFZp781D102
Rabbit
100μL
Antibody
Polyclonal
Human
ImmunohistochemistryĆ¾Western Blot
Unconjugated
CNTN2
Serum
CNTN2
A synthetic peptide from human contactin-2
store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
6900
Videos