missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contactin 2, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen OSC00012W
Code : LD
Additional Details : Weight : 0.01000kg
Description
Reconstitute with 100 ul of distilled water.Specifications
Contactin 2 | |
Polyclonal | |
whole serum with no preservative | |
Q61330, P22063 | |
Axonin-1, Axonal glycoprotein TAG-1, Transient axonal glycoprotein 1, TAX-1, CNTN2, AXT, TAG1, TAX1, MGC157722, DKFZp781D102 | |
Rabbit | |
100μL | |
Antibody | |
Polyclonal | |
Rat |
Western Blot | |
Unconjugated | |
Cntn2 | |
Serum | |
CNTN2 | |
A synthetic peptide from rat contactin-2 | |
avoiding freeze/thaw cycles., store at -20°C, store at 4°C short term. For long term storage | |
Primary | |
21367 |