missing translation for 'onlineSavingsMsg'
Learn More

Contactin 1, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen OSC00026W

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11510463

  • £257.00 / 100ĀµL


Estimated Shipment
15-06-2018
Add to basket

Description

Description

Reconstitute with 100 ul of distilled water.
Specifications

Specifications

Contactin 1
Polyclonal
whole serum with no preservative
P12960
Neural cell surface protein F3, F3cam, contactin 1, CNTN1
Rabbit
100μL
Antibody
Polyclonal
Murine
ImmunohistochemistryĆ¾Western Blot
Unconjugated
Cntn1
Serum
CNTN1
A synthetic peptide from mouse Contactin-1
store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
12805
Videos