Learn More
Complement Factor H Monoclonal Antibody (L20/3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen GAU0200302
Code : LD
Additional Details : Weight : 0.01000kg
Description
GAU 020-03-02 detects factor H in human samples. GAU 020-03-02 has been successfully used in ELISA and Western blot procedures. GAU 018-03-02 can be used as a capture antibody in sandwich ELISA with GAU 020-03-02 as detection antibody. This antibody pair is specific for the ∽150 kDa Factor H protein. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
Complement Factor H Monoclonal antibody specifically detects Complement Factor H in Human samples. It is validated for ELISA, Western Blot.Specifications
Complement Factor H | |
Monoclonal | |
0.91 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P08603 | |
CFH | |
Human factor H | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
L20/3 | |
Unconjugated | |
CFH | |
adrenomedullin binding protein; age-related maculopathy susceptibility 1; AHUS1; AMBP1; ARMD4; ARMS1; beta-1H; beta-1-H-globulin; Cfh; CFHL3; complement component factor H; complement factor H; complement factor H related protein 3A4/5G4; complement factor H, isoform b; complement inhibitory factor H; factor H; factor H-like 1; FH; FHL1; FHL1 antibody; H factor 1; H factor 1 (complement); H factor 2 (complement); HF; HF1; HF2; HGNC:4883; HUS; MGC88246; Mud-1; NOM; platelet complement factor H; protein beta-1-H; RP1-177P10.1; Sas1; Sas-1 | |
Mouse | |
Protein A | |
RUO | |
3075 | |
4° C, store in dark | |
Liquid |