missing translation for 'onlineSavingsMsg'
Learn More

Complement Component C2b Mouse anti-Human, Clone: 269716, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1936

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459045

  • £249.49 / 100µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Complement Component C2b Monoclonal antibody MAB1936 Specifically detects Complement Component C2b in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Complement Component C2b
269716
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Mouse
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects human Complement Component C2b in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) C1r, rhC1s, rhC3r, rhC5a, rhFactor B, or rhFactor I is observed. In Western blots, this antibody recognizes rhC2 and rhC2B.
Human
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
P06681
C2
Mouse myeloma cell line NS0-derived recombinant human Complement Component C2, Ala21-Leu752, Accession # P06681
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
717
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos

For Research Use Only