missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C4 Monoclonal Antibody (KT29), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA172520
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody can be used as a primary antibody for ELISA (1 μg/mL).
Complement C4 Monoclonal antibody specifically detects Complement C4 in Human samples. It is validated for ELISA.Specifications
Complement C4 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P0C0L4, P0C0L5 | |
C4A, C4B | |
Human C4 purified from blood. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
KT29 | |
Unconjugated | |
C4A | |
acidic C4; acidic complement C4; Basic complement C4; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 2; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3; C4; c4 complement; C4-1; C4a; C4a anaphylatoxin; C4A2; C4A3; C4A4; C4A6; C4AD; C4B; C4B_2; C4B1; C4B12; C4B2; C4B3; C4B5; C4b-A; C4b-B; C4BD; C4d-A; C4d-B; C4F; C4S; CH; Chido form of C4; CO4; Complement C4; Complement C4 alpha chain; Complement C4 beta chain; Complement C4 gamma chain; Complement C4-A; Complement C4-A alpha chain; Complement C4-B; Complement C4-B alpha chain; complement C4B1a; complement component 4 (within H-2S); complement component 4, gene 1; complement component 4A (Rodgers blood group); complement component 4B (Chido blood group); complement component 4B (Childo blood group); CPAMD2; CPAMD3; DADB-112B14.11; DAMC-258G8.6; FLJ60561; MGC164979; MHC class III region complement; RG; Rodgers form of C4; sex-limited protein; Slp; Ss | |
Mouse | |
Protein A | |
RUO | |
100293534, 720, 721 | |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
Liquid |