Learn More
Complement C3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA140288
Code : LD
Additional Details : Weight : 0.01000kg
Description
PA1-40288 detects C3 from mouse samples. PA1-40288 has been successfully used in immunohistochemistry (frozen), immunohistochemistry (paraffin), immunofluorescence and Western blot applications. The antibody works well in both PBS and in TBS. The PA1-40288 imunogen is mouse C3. Positive control: Kidney, Spleen.
Complement C3 Polyclonal antibody specifically detects Complement C3 in Human, Mouse samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
Complement C3 | |
Polyclonal | |
Unconjugated | |
C3 | |
Acylation stimulating protein; acylation-stimulating protein cleavage product; AHUS5; AI255234; ARMD9; ASP; C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a; C3a anaphylatoxin; C3adesArg; C3b; C3bc; C3-beta-c; C3d; complement C3; Complement C3 alpha chain; Complement C3 beta chain; complement C3 protein (GPC3) precursor; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component 3; complement component C3; complement component C3 alpha-chain; complement component C3a; complement component C3b; complement component C3d; CPAMD1; ENCF-1; ENCF-2; epididymis secretory sperm binding protein Li 62p; HEL-S-62p; HSE-MSF; I79_008227; LOC100060539; Neutrophil chemotactic factor-1; Neutrophil chemotactic factor-2; Plp; prepro-C3; Unknown (protein for MGC:134562) | |
Rabbit | |
Protein G | |
RUO | |
12266, 718 | |
4° C | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 0.1% BSA and 0.02% sodium azide | |
P01024, P01027 | |
C3 | |
Murine C3. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |