Learn More
Collagen II Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA126206
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: chondrocyte tissue. PA1-26206 is expected to cross react with most mammals due to sequence homology. This antibody shows negligable cross reactivity with the Collagens 1, 3, 4, 5, and 6.
Collagen II Polyclonal antibody specifically detects Collagen II in Bovine, Human, Mouse, Ovine, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
Collagen II | |
Polyclonal | |
Unconjugated | |
Col2a1 | |
alpha 1 type II procollagen; alpha 1 type IIA collagen; alpha 1 type IIA collagen precursor; alpha collagen type II; alpha-1 type II collagen; ANFH; AOM; arthroophthalmopathy, progressive (Stickler syndrome); cartilage collagen; CB11; CG2A1A; CGA; Chondrocalcin; COL11A3; Col2; Col2a; col2a1; Col2a-1; col2a1.L; col2a1a; col2a1-a; col2a1-b; Collagen 2; Collagen alpha-1(II) chain; collagen II, alpha-1 polypeptide; collagen type II (Col2A1) gene, enhancer region; collagen type II alpha 1 chain; collagen type IIA; collagen, type II, alpha 1; collagen, type II, alpha 1 (primary osteoarthritis, spondyloepiphyseal dysplasia, congenital); collagen, type II, alpha 1 L homeolog; COLLII; cyanogen bromide; Del1; disproportionate micromelia; Dmm; Lpk; M100856; MGC131516; pro alpha1 type II collagen; pro-alpha 1(II); pro-alpha-1 type II collagen; procollagen alpha 1(II); Procollagen II alpha 1; procollagen, type II, alpha 1; Rgsc856; SEDC; STL1; type II collagen; type II collagen cyanogen bromide fra | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100196905, 1280, 12824, 25412, 407142 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1.03 mg/mL | |
0.125M sodium borate with 0.075M NaCl, 5mM EDTA and 0.01% sodium azide; pH 8.0 | |
P02458, P02459, P05539, P28481 | |
Col2a1 | |
Collagen type II from adult human knee cartilage and bovine nasal cartilage. | |
100 μg | |
Primary | |
Bovine, Human, Mouse, Ovine, Rat | |
Antibody | |
IgG |