Learn More
COCO Rat anti-Mouse, Clone: 340531, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3356
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13450026
  • £277.83 / 100µg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
COCO Monoclonal antibody MAB3356 Specifically detects COCO in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
COCO/DAND5
340531
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
DAND5
E. coli-derived recombinant mouse COCO Arg24-Leu185 Accession # Q76LW6
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
199699
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q76LW6
CER2CRL2, Cerberus 2, Cerberus like-2, cerberus-like 2, Cerberus-like protein 2, cerl-2, CERL2, CKTSF1B3, CKTSF1B3MGC126849, COCO, Cysteine knot superfamily 1, BMP antagonist 3, DAN domain family member 5, DAN domain family, member 5, DAND5, Dante, DTE, FLJ38607, GREM3, GREM3cerberus 2, Gremlin-3, SP1
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse COCO in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) COCO, rhDAN, recombinant mouse (rm) Gremlin, rmCerberus, and recombinant chicken Caronte is observed.