Learn More
COCO Mouse anti-Human, Clone: 322209, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB30471
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439855
  • £337.93 / 500µg
Estimated Shipment
27-06-2019
Description
Description
COCO Monoclonal antibody MAB30471 Specifically detects COCO in Human samples. It is validated for ELISA Capture
Specifications
Specifications
COCO/DAND5
322209
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Q8N907
CER2CRL2, Cerberus 2, Cerberus like-2, cerberus-like 2, Cerberus-like protein 2, cerl-2, CERL2, CKTSF1B3, CKTSF1B3MGC126849, COCO, Cysteine knot superfamily 1, BMP antagonist 3, DAN domain family member 5, DAN domain family, member 5, DAND5, Dante, DTE, FLJ38607, GREM3, GREM3cerberus 2, Gremlin-3, SP1
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human COCO in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) Activin A, rhBMP-4, rhDAN, or recombinant mouse COCO is observed.
Human
ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
DAND5
E. coli-derived recombinant human COCO Arg23-Ala189 Accession # Q8N907
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
199699
Reconstitute at 0.5 mg/mL in sterile PBS.