Learn More
Coagulation Factor XIV/Protein C Mouse anti-Human, Clone: 390226, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3349
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13430026
  • £286.65 / 100µg
Estimated Shipment date
from Supplier 31-03-2020
Description
Description
Coagulation Factor XIV/Protein C Monoclonal antibody MAB3349 Specifically detects Coagulation Factor XIV/Protein C in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Coagulation Factor XIV/Protein C
390226
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
PROC
Mouse myeloma cell line NS0-derived recombinant human Coagulation Factor XIV/Protein C Ala43-Pro461 Accession # P04070
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5624
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
P04070
Anticoagulant protein C, APC, Autoprothrombin IIA, Blood coagulation factor XIV, EC 3.4.21, EC 3.4.21.69, PC, PROC, PROC1, Protein C, protein C (inactivator of coagulation factors Va and VIIIa), vitamin K-dependent protein C
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Coagulation Factor XIV/Protein C in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Factor VII, rhFactor X, rhFactor XI, rhThrombin, rhUPA, rhHGFA, rhKallikrein 1, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12, 13, 14, or 15 is observed.