Learn More
Coagulation Factor X Mouse anti-Human, Clone: 156106, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1063
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13468295
  • £417.48 / 500µg
Estimated Shipment date
from Supplier 24-03-2020
Description
Description
Coagulation Factor X Monoclonal antibody MAB1063 Specifically detects Coagulation Factor X in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Coagulation Factor X
156106
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
F10
Mouse myeloma cell line NS0-derived recombinant human Coagulation Factor X Leu24-Lys488 (predicted) Accession # P00742
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2159
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
P00742
Cf10, coagulation factor X, EC 3.4.21, EC 3.4.21.6, F10, factor Xa, FX, FXa, prothrombinase, Stuart Factor, Stuart-Prower factor
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Coagulation Factor X in direct ELISAs and Western blots. It recognizes both Factor X and Factor Xa.