Learn More
Coagulation Factor III/Tissue Factor Rat anti-Mouse, Clone: 355220, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3178
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13459935
  • £249.49 / 100µg
Estimated Shipment
07-03-2019
Add to basket
Description
Description
Coagulation Factor III/Tissue Factor Monoclonal antibody MAB3178 Specifically detects Coagulation Factor III/Tissue Factor in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Coagulation Factor III/Tissue Factor
355220
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
F3
Mouse myeloma cell line NS0-derived recombinant mouse Coagulation Factor III Ala29-Glu251 Accession # P20352
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2152
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P20352
CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), F3, FLJ17960, TF, TFA, Thromboplastin, tissue factor
Rat
IgG2c
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Coagulation Factor III/Tissue Factor in direct ELISAs and Western blots.