Learn More
Coagulation Factor III/Tissue Factor Mouse anti-Human, Clone: 323514, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2339-100
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13469365
  • £311.64 / 100µg
Estimated Shipment date
from Supplier 24-03-2020
Description
Description
Coagulation Factor III/Tissue Factor Monoclonal antibody MAB2339100 Specifically detects Coagulation Factor III/Tissue Factor in Human samples. It is validated for ELISA Capture, Immunoprecipitation, Western Blotting
Specifications
Specifications
Coagulation Factor III/Tissue Factor
323514
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
F3
Mouse myeloma cell line NS0-derived recombinant human Coagulation Factor III Gly34-Glu251 Accession # P13726
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2152
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation, ELISA
LYOPH
Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
P13726
CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), F3, FLJ17960, TF, TFA, Thromboplastin, tissue factor
Mouse
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human coagulation factor III/Tissue Factor in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse Tissue Factor is observed.