missing translation for 'onlineSavingsMsg'
Learn More

Pro Coagulation Factor II/Thrombin Mouse anti-Human, Clone: 200710, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1473

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458665

  • £337.93 / 500µg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

Pro Coagulation Factor II/Thrombin Monoclonal antibody MAB1473 Specifically detects Pro Coagulation Factor II/Thrombin in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Coagulation Factor II/Thrombin
200710
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
F2
Mouse myeloma cell line NS0-derived recombinant human Pro Coagulation Factor II/Thrombin Ala44-Glu622 Accession # NP_000497
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2147
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
NP_000497
Coagulation Factor II, coagulation factor II (thrombin) receptor-like 2, Coagulation factor II receptor-like 2, Coagulation factor II receptor-like 2 (protease-actovated receptor 3), F2, PAR-3, PAR3proteinase-activated receptor 3, protease-activated receptor 3, proteinase-activated receptor-3, PT, serine protease, Thrombin, Thrombin receptor-like 2
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Pro Coagulation Factor II/Thrombin in direct ELISAs and Western blots. It recognizes only Pro Coagulation Factor II/Thrombin, not active Thrombin. In Western blots, less than 5% cross-reactivity with recombinant human (rh) Factor X, rhKallikrein 1, 3, 5, and 11 is observed.
Human
Videos

For Research Use Only