Learn More
CNP Rat anti-Human, Clone: 462608, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB31271
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13469905
  • £286.65 / 100µg
Estimated Shipment date
from Supplier 27-03-2020
Description
Description
CNP Monoclonal antibody MAB31271 Specifically detects CNP in Human samples. It is validated for Western Blotting
Specifications
Specifications
CNP/NPPC
462608
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
NPPC
E. coli-derived recombinant human CNP Lys24-Cys126 Accession # P23582
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4880
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P23582
CNP, CNP2, C-type natriuretic peptide, natriuretic peptide C, natriuretic peptide precursor C, NPPC
Rat
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human pro-CNP and mature CNP53 and CNP29 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) ANP or rhBNP is observed.