Learn More
Clostridium perfringens Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA185310
Code : LD
Additional Details : Weight : 0.01000kg
Description
Rabbit anti Clostridium perfringens antibody recognizes Clostridium perfringens, a Gram positive bacterium widespread in the environment. In humans it can be pathogenic, causing gastrointestinal illness and gangrene.
Clostridium perfringens Polyclonal antibody specifically detects Clostridium perfringens in Bacteria samples. It is validated for ELISA.Specifications
Clostridium perfringens | |
Polyclonal | |
Unconjugated | |
C. perfringens | |
Clostridium perfringens ATCC 13124. | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA | |
5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Rabbit | |
Protein G | |
RUO | |
Bacteria | |
Antibody | |
IgG |