missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLA, Rat anti-Human, Clone: HECA-452, BV421, BD
Brand: BD Biosciences 563961
Additional Details : Weight : 0.01000kg
Description
Host Species: Rat
Clone: HECA-452
Isotype: IgM, κ
Species Reactivity: Hu
Immunogen: Lymphocyte-depleted Stromal Preparation of Tonsil
Flow cytometry
Specifications
CLA | |
HECA-452 | |
Rat | |
Affinity Purified | |
RUO | |
Human |
Monoclonal | |
Brilliant Violet 421 | |
Lymphocyte-depleted Stromal Preparation of Tonsil | |
100 Tests | |
Primary |