Learn More
CHL-1/L1CAM-2 Rat anti-Mouse, Clone: 316316, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2147
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13469215
  • £286.65 / 100µg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
CHL-1/L1CAM-2 Monoclonal antibody MAB2147 Specifically detects CHL-1/L1CAM-2 in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
CHL-1/L1CAM-2
316316
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CHL1
Mouse myeloma cell line NS0-derived recombinant mouse CHL-1/L1CAM-2 Ala25-Gln1043 (Leu227-Gln242 del & Ala243Ser) Accession # BAC30699
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
10752
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
BAC30699
CALL, CALLClose homolog of L1, cell adhesion molecule with homology to L1CAM (close homolog of L1), cell adhesion molecule with homology to L1CAM (close homologue of L1), CHL1, FLJ44930, L1CAM-2, L1CAM2L1 cell adhesion molecule 2, MGC132578, neural cell adhesion molecule L1-like protein
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse CHL-1/L1CAM-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CHL-1 is observed.