Learn More
RGM-A Goat anti-Chicken, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF2010
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13487284
  • £362.00 / 100µg
Estimated Shipment
07-03-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
RGM-A Polyclonal antibody AF2010 Specifically detects RGM-A in Chicken samples. It is validated for Western Blotting
Specifications
Specifications
RGM-A
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant chicken&No.160;RGM-A, Trp30-Asn404 (Asn404Asp), Accession No. Q8JG54
100μg
Primary
Chicken
Unconjugated
Q8JG54
Goat