Learn More
Goat anti-Chicken IgY (H+L) Secondary Antibody, Qdot™ 655, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen Q14421MP
Code : 50
Additional Details : Weight : 0.50000kg
Description
Qdot nanocrystals are composed of semi-conductor material to generate a fluorescent particle which is exceptionally bright and does not photobleach. Qdot nanocrystals paired with the correct optical filters are as much as 50 times brighter than traditional organic dyes.
Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Chicken IgY (H+L) | |
Polyclonal | |
Qdot 655 | |
Goat | |
purified | |
RUO | |
Chicken | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot | |
1 μM | |
0.05M borate with 1M betaine and 0.05% sodium azide; pH 8.3 | |
Chicken IgY (H+L) | |
200 μL | |
Secondary | |
4° C, store in dark | |
IgG |