missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD99 Monoclonal Antibody (MEM-131), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119578
Code : LD
Additional Details : Weight : 0.01000kg
Description
CD99 Monoclonal antibody specifically detects CD99 in Human samples. It is validated for Flow Cytometry.Specifications
CD99 | |
Monoclonal | |
FITC | |
CD99 | |
1110061M03Rik; 12E7; 2410026K10Rik; antigen identified by monoclonal 12E7, Y homolog; antigen identified by monoclonal antibodies 12E7, F21 and O13; Cd99; CD99 antigen; CD99 molecule; cell surface antigen 12E7; cell surface antigen HBA-71; cell surface antigen O13; D4; E2 antigen; HBA71; MIC2; MIC2 (monoclonal antibody 12E7); MIC2X; MIC2Y; MSK5X; paired immunoglobin-like type 2 receptor-ligand; pilr-1; Pilrl; PILR-L; Protein MIC2; surface antigen MIC2; T-cell surface glycoprotein E2 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
4267 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MEM-131 | |
TBS with 0.2% BSA and 15mM sodium azide; pH 8.0 | |
P14209 | |
CD99 | |
HPB-ALL human peripheral blood leukeima T-cell line. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgM |