CD97 Polyclonal Antibody, Invitrogen™

CD97 Antibody, PA5-33413, from Invitrogen™. Species Reactivity: Human; Applications: Immunocytochemistry, Immunohistochemistry (Paraffin).

Overview
Brand: Invitrogen

Manufacturer Part Number: PA5-33413

UNSPSC: 12352203

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 15374491

Quantity Price
1 £ 314.0 / 50µg
Estimated Shipment date
from Supplier 10-04-2017
Add to basket


Description and Specification

Specification

Formulation 50μg purified antibody (1mg/mL) in PBS and < 0.1% sodium azide
Gene ID (Entrez) 976
Cross Reactivity Human
Antigen CD97
Storage Requirements Store at 2-8°C for up to 1 month. For long term storage store at -20°C
Isotype IgG
Immunogen Synthetic 19 amino acid peptide from N-terminal extracellular domain of human CD97.
Primary or Secondary Primary
Format Purified
Gene Accession No. P48960
Conjugate Unlabeled
Host Species Rabbit
Species Reactivity Human
Monoclonal or Polyclonal Polyclonal
Quantity 50μg
Gene Symbols CD97
Applications Immunocytochemistry
Applications Immunohistochemistry (Paraffin)
Gene Alias CD97, CD97 molecule, Leukocyte antigen CD97, TM7LN1, CD97 antigen

CD97 is a 75-85kDa monomeric glycoprotein that is present on the surface of most activated leucocytes. It is a receptor involved in both cell adhesion and signaling. CD97 is a member of the G protein coupled receptor family. It is expressed abundantly in cells of hematopoietic origin and is markedly upregulated on activated T and B cells. ESTs have been isolated from a wide variety of tissue libraries.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Synthetic 19 amino acid peptide from N-terminal extracellular domain of human CD97.
Formulation: 50 ug purified antibody (1mg/mL) in PBS and < 0.1% sodium azide
Synonyms: CD97, CD97 molecule, Leukocyte antigen CD97, TM7LN1, CD97 antigen
Uniprot Accession: P48960
Entrez GeneID: 976
Gene Symbol: CD97
Percent identity with other species by BLAST analysis: Human (100%) Monkey (95%).