missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD97 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533414
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human (100%) Monkey (94%) Marmoset (88%).
CD97 Polyclonal antibody specifically detects CD97 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
CD97 | |
Polyclonal | |
Unconjugated | |
ADGRE5 | |
AA409984; Adgre5; adhesion G protein-coupled receptor E5; Cd97; CD97 antigen; CD97 antigen subunit alpha; CD97 antigen subunit beta; CD97 molecule; EGF-TM7 receptor; leukocyte antigen CD97; seven transmembrane helix receptor; seven-span transmembrane protein; seven-transmembrane, heterodimeric receptor associated with inflammation; TM7LN1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
976 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P48960 | |
ADGRE5, CD97 | |
Synthetic 17 amino acid peptide from N-terminal extracellular domain of human CD97. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |