Learn More
CD94 Monoclonal Antibody (15F.18D1), FITC, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA517897
142.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA5-17897 targets CD94/KLRD1 in ELISA, FACS, and IF applications and shows reactivity with Mouse samples. The MA5-17897 immunogen is mouse T cell clone AE7 cells.
CD94 Monoclonal antibody specifically detects CD94 in Mouse samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry.Specifications
CD94 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
O54707 | |
KLRD1 | |
Murine T cell clone AE7 cells | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 κ |
ELISA, Flow Cytometry, Immunocytochemistry | |
15F.18D1 | |
FITC | |
KLRD1 | |
Cd94; CD94 antigen; CD94 antigen (located within the rat natural killer gene complex); killer cell lectin like receptor D1; Killer cell lectin-like receptor subfamily D member 1; killer cell lectin-like receptor subfamily D, member 1; killer cell lectin-like receptor, subfamily D, member 1; KLRD1; KP43; natural killer cell receptor; natural killer cells antigen CD94; NK cell receptor | |
Rat | |
Protein G | |
RUO | |
16643 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |