Learn More
CD90/Thy1 Mouse anti-Human, Clone: Thy-1A1, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2067
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13429145
  • £311.64 / 100µg
Estimated Shipment date
from Supplier 06-04-2020
Description
Description
CD90/Thy1 Monoclonal antibody MAB2067 Specifically detects CD90/Thy1 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
CD90/Thy1
Thy-1A1
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD90, CD90 antigen, CDw90, FLJ33325, Thy1, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen
Mouse
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human CD90/Thy1.
Human
Flow Cytometry, CyTOF
LYOPH
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Purified
THY1
Human CD90/Thy1
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
7070
Reconstitute at 0.5 mg/mL in sterile PBS.