Learn More
CD90 Mouse anti-Human, APC, Clone: 5E10., BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 561971
Additional Details : Weight : 0.01000kg
Description
The 5E10 monoclonal antibody specifically binds to human CD90 which is also known as Thy-1. CD90 is a 25-35 kDa glycophosphatidylinositol-anchored membrane glycoprotein of the Ig superfamily that is expressed on 1-4% of human fetal liver cells, cord blood cells, and bone marrow cells. The anti-CD90 antibody binds to a subset of immature CD34+ cells and a distinct subset of mature CD34- cells that are CD3+CD4+. The CD90+CD34+ population is highly enriched for cells capable of long-term culture. The anti-CD90 antibody is useful for enriching high proliferative potential colony-forming cells (HIPP-CFC) that are primative progenitor cells.
Specifications
CD90 | |
Monoclonal | |
0.2mg/mL | |
THY1; Thy-1 antigen; Thy-1 membrane glycoprotein | |
Mouse | |
Affinity Purified | |
RUO | |
Baboon, Canine, Cynomolgus, Human, Porcine, Rhesus Monkey | |
IgG1 κ |
Flow Cytometry | |
5E10. | |
APC | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human HEL Cell Line | |
25 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |