missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD9 Monoclonal Antibody (IVA50), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119301
309.70 GBP valid until 2024-03-29
Use promo code "21653" to get your promotional price.
Code : LD
Additional Details : Weight : 0.01000kg
Description
CD9 Monoclonal antibody specifically detects CD9 in Bovine, Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Western Blot.Specifications
CD9 | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P21926, P30932 | |
CD9 | |
Bovine thrombocytes. | |
100 μg | |
Primary | |
Bovine, Human | |
Antibody | |
IgG2 |
Flow Cytometry, Immunoprecipitation, Western Blot | |
IVA50 | |
Unconjugated | |
CD9 | |
5H9; 5H9 antigen; antigen CD9; BA2; BA-2/p24 antigen; BTCC-1; CD9; CD9 antigen; CD9 antigen (p24); CD9 molecule; cell growth-inhibiting gene 2 protein; DRAP-27; GIG2; Leukocyte antigen MIC3; MIC3; motility related protein-1; Motility-related protein; MRP-1; p24; sCD 9; sCD9; soluble CD 9; soluble CD9; Tetraspanin29; Tetraspanin-29; transmembrane protein CD9; TSPAN29; Tspan-29 | |
Mouse | |
Protein A | |
RUO | |
280746, 928 | |
4° C, do not freeze | |
Liquid |