Learn More
CD9 Rat anti-Mouse, Clone: 479608, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB5218
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13460856
  • £273.25 / 100µg
Estimated Shipment
07-03-2019
Add to basket
Description
Description
CD9 Monoclonal antibody MAB5218 Specifically detects CD9 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry
Specifications
Specifications
CD9
479608
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CD9
NS0 mouse myeloma cell line transfected with mouse CD9 Pro2-Val226 Accession # P40240
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
928
Reconstitute at 0.5 mg/mL in sterile PBS.
Flow Cytometry, Immunohistochemistry, CyTOF
LYOPH
Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready
P40240
BA2, BA-2/p24 antigen, BTCC-1, CD9, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24)
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Stains mouse CD9-transfected cells but not irrelevant transfecants.