missing translation for 'onlineSavingsMsg'
Learn More

anti-CD8; Clone: RPA-T8; BD

Brand:  BD Biosciences 563256

Code : NEW

Additional Details : Weight : 0.00100kg

Product Code. 15725978

  • £284.00 / 50 tests


Estimated Shipment date
from Supplier 24-01-2019
Add to basket

Description

Description

Host Species: Mouse
Clone RPA-T8
Isotype IgG1, κ
Species Reactivity Hu, Rh, Cyn, Bb

Flow Cytometry

Specifications

Specifications

CD8
Brilliant Violet 510
Murine
50 Tests
Primary
Baboon, Cynomolgus, Human, Rhesus
RPA-T8
Affinity Purified
IgG1
RUO
Monoclonal
Videos