Learn More
CD8 beta Monoclonal Antibody (3.4.1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA170040
147.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-70040 detects CD8b in rat samples. MA1-70040 has been successfully used in flow cytometry procedures. The MA1-70040 immunogen is rat/mouse T cell hybrids expressing CD8.
CD8 beta Monoclonal antibody specifically detects CD8 beta in Rat samples. It is validated for Flow Cytometry.Specifications
CD8 beta | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P05541 | |
Cd8b | |
Rat/mouse T cell hybrids expressing CD8. | |
250 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry | |
3.4.1 | |
Unconjugated | |
Cd8b | |
CD8; CD8 antigen 37 kDa chain; CD8 antigen beta polypeptide; CD8 antigen beta polypeptide precursor; CD8 antigen beta-chain; CD8 antigen, beta chain; CD8 antigen, beta chain 1; CD8 antigen, beta polypeptide 1 (p37); CD8 antigen, beta-chain; CD8 beta; CD8 beta chain; CD8a; CD8alpha; CD8B; CD8b antigen; CD8b molecule; CD8b molecule pseudogene; CD8B1; CD8beta; CD8BP; LEU2; Leu-2; LY3; Ly-3; Ly-C; Lymphocyte antigen 3; Lyt3; Lyt-3; MAL; OX-8 membrane antigen; p32; P37; T lymphocyte surface glycoprotein beta chain; T-cell membrane glycoprotein Ly-3; T-cell surface glycoprotein; T-cell surface glycoprotein CD8 beta chain; T-cell surface glycoprotein Lyt-3 | |
Mouse | |
Protein G | |
RUO | |
24931 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |