Learn More
CD8 alpha Monoclonal Antibody (OX-8), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA180658
Code : LD
Additional Details : Weight : 0.01000kg
Description
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Rat CD8α, clone MRC OX-8, recognizes the rat CD8 alpha cell surface antigen, expressed by a subset of T lymphocytes, most thymocytes and the majority of NK cells.
CD8 alpha Monoclonal antibody specifically detects CD8 alpha in Rat samples. It is validated for Flow Cytometry.Specifications
CD8 alpha | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA and 0.09% sodium azide; pH 7.4 | |
P05541, P07725 | |
Cd8a, CD8B | |
Rat thymocyte membrane glycoproteins. | |
100 Tests | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry | |
OX-8 | |
PE | |
CD8B | |
BB154331; CD8; CD8 alpha; CD8 alpha chain; CD8 alpha chain precursor; CD8 antigen; CD8 antigen 32 kDa chain; CD8 antigen 37 kDa chain; CD8 antigen alpha polypeptide; CD8 antigen alpha protein; CD8 antigen alpha protein precursor; CD8 antigen alpha-chain; CD8 antigen beta polypeptide; CD8 antigen beta polypeptide precursor; CD8 antigen beta-chain; CD8 antigen, alpha chain; CD8 antigen, alpha polypeptide; CD8 antigen, alpha polypeptide (p32); CD8 antigen, alpha-chain; CD8 antigen, beta chain; CD8 antigen, beta chain 1; CD8 antigen, beta polypeptide; CD8 antigen, beta polypeptide 1 (p37); CD8 antigen, beta-chain; CD8 beta; CD8 beta chain; CD8 beta-2; CD8a; CD8A antigen alpha; CD8a molecule; CD8A; T-cell surface glycoprotein; CD8alpha; CD8B; CD8b antigen; CD8b molecule; CD8b molecule pseudogene; Cd8b1; CD8beta; CD8BP; fCD8; LEU2; Leu-2; Leu2 T-lymphocyte antigen; leu-2a; Ly-2; LY3; Ly-3; Ly-35; Ly-B; Ly-C; Lymphocyte antigen 3; Lyt2; Lyt-2; Lyt-2.1 lymphocyte differentiation antigen (AA at | |
Mouse | |
Protein G | |
RUO | |
24930, 24931 | |
4° C, store in dark, DO NOT FREEZE! | |
Lyophilized |