Learn More
CD72 Rat anti-Mouse, Clone: 164726, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1279
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13468515
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 03-04-2020
Description
Description
CD72 Monoclonal antibody MAB1279 Specifically detects CD72 in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
CD72
164726
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CD72
Mouse myeloma cell line NS0-derived recombinant mouse CD72 Arg117-Asn354 Accession # P21855
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
971
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P21855
CD72, CD72 antigenB-cell differentiation antigen CD72, CD72 molecule, CD72b, Ly-19.2, Ly-32.2, Lyb-2, LYB2lyb-2
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse CD72 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human CD72 is observed.