missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD7 Monoclonal Antibody (MEM-186), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119080
Code : LD
Additional Details : Weight : 0.01000kg
Description
Western Blot: non-reducing conditions.
CD7 Monoclonal antibody specifically detects CD7 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
CD7 | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P09564 | |
CD7 | |
Human acute myelogenous leukaemia cell line KG-1. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
MEM-186 | |
Unconjugated | |
CD7 | |
Cd7; CD7 antigen; CD7 antigen (p41); Cd7 molecule; GP40; LEU-9; p41 protein; T-cell antigen CD7; T-cell leukemia antigen; T-cell surface antigen Leu-9; Tp40; TP41 | |
Mouse | |
Protein A | |
RUO | |
924 | |
4° C, do not freeze | |
Liquid |