missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEA Monoclonal Antibody (26/3/13), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA117759
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody recognizes native human CEACAM5 protein expressed in transfected mammalian cells.
CEA Monoclonal antibody specifically detects CEA in Human samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Western Blot.Specifications
CEA | |
Monoclonal | |
2 mg/mL | |
PBS with no preservative; pH 7.2 | |
P06731 | |
Ceacam5 | |
extracted protein of human CEACAM5 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Western Blot | |
26/3/13 | |
Unconjugated | |
Ceacam5 | |
1600029H12Rik; Carcinoembryonic antigen; carcinoembryonic antigen related cell adhesion molecule 5; carcinoembryonic antigen-related cell adhesion molecule 5; CD66e; CEA; CEACAM5; Meconium antigen 100; OTTHUMP00000199033; pregnancy-specific glycoprotein 30; Psg30; sCD66e; soluble CD66e | |
Mouse | |
Protein G | |
RUO | |
1048 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |