Learn More
CD66 Mouse anti-Human, FITC, Clone: B1.1/CD66, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 551479
Additional Details : Weight : 0.02000kg
Description
The B1.1 monoclonal antibody binds to several glycosylphosphatidylinositol-anchored glycoproteins present on granulocytes and epithelial cells. As reported in the Sixth International Workshop and Conference on Human Leucocyte Differentiation Antigens, the B1.1 antibody recognized CD66a (CEACAM1), CD66c (CEACAM6), CD66d (CEACAM3) and CD66e (. These molecules exhibit both homophilic and heterophilic adhesion. CEA family members may be involved in transmembrane signaling and activation of neutrophils.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
CD66 | |
Monoclonal | |
FITC | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human Breast Carcinoma Cells | |
100 Tests | |
Immunology | |
Human | |
IgG2a κ |
Flow Cytometry | |
B1.1/CD66 | |
CEA, carcinoembryonic antigen, CD66a, CD66c, CD66d, CD66e | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.