missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD64 Monoclonal Antibody (290322), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA523993
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In Western blots, no cross-reactivity with recombinant human (rh) Fc gamma RIA, rhFc gamma RIIB, or rhFc gamma RIIIB is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
CD64 Monoclonal antibody specifically detects CD64 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD64 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P26151 | |
Fcgr1 | |
NS0-derived recombinant mouse Fc gamma RI extracellular domain Glu25-Pro297 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry | |
290322 | |
Unconjugated | |
Fcgr1 | |
AI323638; AV092959; CD64; CD64 antigen; CD64A; Fc fragment of IgG high affinity Ib receptor; Fc fragment of IgG receptor Ia; Fc fragment of IgG, high affinity Ia, receptor (CD64); Fc fragment of IgG, high affinity Ia, receptor for (CD64); Fc fragment of IgG, high affinity Ib, receptor for (CD64); Fc gamma receptor; Fc gamma receptor Ia; Fc receptor, IgG, high affinity I; Fcg1; Fc-gamma receptor 1; Fc-gamma receptor I A1; Fc-gamma RI; Fc-gamma RIA; FcgammaRI; FcgammaRIa; Fcgr1; FCGR1A; Fcgr1b; FcgRI; FCRI; FLJ18345; high affinity IgG Fc receptor, subunit beta; high affinity immunoglobulin gamma Fc receptor I; high-affinity immunoglobulin gamma Fc receptor I; IGFR1; IgG Fc receptor I; IgG high affinity Fc receptor; IGGHAFC; RP11-196G18.2 | |
Rat | |
Protein A/G | |
RUO | |
14129 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |