Learn More
CD6 Monoclonal Antibody (6B9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517047
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA5-17047 targets CD6 in IHC, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17047 immunogen is purified recombinant fragment of human CD6 (amino acids: 472-668) expressed in E. Coli. MA5-17047 detects CD6 which has a predicted molecular weight of approximately 71.8kDa.
CD6 Monoclonal antibody specifically detects CD6 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot.Specifications
CD6 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P30203 | |
CD6 | |
Purified recombinant fragment of human CD6 (amino acids: 472-668) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
6B9 | |
Unconjugated | |
CD6 | |
Cd6; CD6 antigen; CD6 antigen, Tp120; Cd6 molecule; FLJ44171; OX52; Soluble CD6; T12; T-cell differentiation antigen CD6; TP120 | |
Mouse | |
Protein G | |
RUO | |
923 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |