Learn More
ICAM-3 Monoclonal Antibody (MEM-04), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119030
Code : LD
Additional Details : Weight : 0.01000kg
Description
Low endotoxin azide free phosphate buffered saline (PBS) solution, 0.2 μm filter sterilized. Endotoxin level is less than 0.01 EU/μg of the protein, as determined by the LAL test. Functional Application: Blocking.
ICAM-3 Monoclonal antibody specifically detects ICAM-3 in Human samples. It is validated for Flow Cytometry, Functional Assay, Immunohistochemistry (Paraffin).Specifications
ICAM-3 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
P32942 | |
ICAM3 | |
Leukocytes from a patient suffering from LGL type leukemia. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Flow Cytometry, Functional Assay, Immunohistochemistry (Paraffin) | |
MEM-04 | |
Unconjugated | |
ICAM3 | |
CD50; CDw50; ICAM3; ICAM-3; ICAM-R; Intercellular adhesion molecule 3; intercellular adhesion molecule-3 | |
Mouse | |
Protein A | |
RUO | |
3385 | |
4° C, do not freeze | |
Liquid |