Learn More
CD49f Monoclonal Antibody (NKI-GoH3), PE, Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen MA516885
Code : NEW
Additional Details : Weight : 0.01000kg
Beschreibung
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
CD49f Monoclonal antibody specifically detects CD49f in Canine, Cynomolgus Monkey, Human, Mouse, Ovine, Porcine samples. It is validated for Flow Cytometry.Spezifikation
CD49f | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
P23229, Q61739 | |
Itga6 | |
BALB/c mouse mammary tumour cells. | |
100 Tests | |
Primary | |
Canine, Cynomolgus Monkey, Human, Mouse, Ovine, Porcine | |
Antibody | |
IgG2a |
Flow Cytometry | |
NKI-GoH3 | |
PE | |
Itga6 | |
5033401O05Rik; AI115430; Alpha6p; CD49 antigen-like family member F; CD49f; DKFZp686J01244; FLJ18737; integrin alpha 6; integrin alpha-6; Integrin alpha-6 heavy chain; Integrin alpha-6 light chain; integrin alpha6B; integrin subunit alpha 6; integrin, alpha 6; Itga6; ITGA6B; LOW QUALITY PROTEIN: integrin alpha-6; Processed integrin alpha-6; VLA-6 | |
Rat | |
Protein G | |
RUO | |
100158200, 100856563, 101113656, 101925271, 16403, 3655 | |
4° C, store in dark, DO NOT FREEZE! | |
Lyophilized |