Learn More
CD49f Monoclonal Antibody (NKI-GoH3), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA516884
Code : NEW
Additional Details : Weight : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. A suggested positive control for immunohistochemical applications is human tonsil.
CD49f Monoclonal antibody specifically detects CD49f in Canine, Cynomolgus Monkey, Human, Mouse, Ovine, Porcine samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation.Specifications
CD49f | |
Monoclonal | |
1 mg/mL | |
PBS with <0.1% sodium azide | |
P23229, Q61739 | |
Itga6 | |
BALB/c mouse mammary tumour cells | |
100 μg | |
Primary | |
Canine, Cynomolgus Monkey, Human, Mouse, Ovine, Porcine | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation | |
NKI-GoH3 | |
Unconjugated | |
Itga6 | |
5033401O05Rik; AI115430; Alpha6p; CD49 antigen-like family member F; CD49f; DKFZp686J01244; FLJ18737; integrin alpha 6; integrin alpha-6; Integrin alpha-6 heavy chain; Integrin alpha-6 light chain; integrin alpha6B; integrin subunit alpha 6; integrin, alpha 6; Itga6; ITGA6B; LOW QUALITY PROTEIN: integrin alpha-6; Processed integrin alpha-6; VLA-6 | |
Rat | |
Protein G | |
RUO | |
100158200, 100856563, 101113656, 101925271, 16403, 3655 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |