Learn More
ITGA4 Monoclonal Antibody (44H6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516515
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Overnight incubation with clone 44H6 is recommended for optimal results. The epitope recognized by this antibody is reported to be sensitive to formaldehyde fixation and tissue processing. We recommend the use of acetone fixation for frozen sections. A suggested positive control for immunohistochemical applications is human tonsil. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
ITGA4 Monoclonal antibody specifically detects ITGA4 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen).Specifications
ITGA4 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P13612 | |
ITGA4 | |
HOON pre-B leukaemia cell line | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen) | |
44H6 | |
Unconjugated | |
ITGA4 | |
269C wild type; alpha 4 subunit of VLA-4 receptor; antigen CD49D, alpha-4 subunit of VLA-4 receptor; CD49 antigen-like family member D; CD49d; IA4; integrin alpha 4; integrin alpha-4; integrin alpha4B; Integrin alpha-IV; integrin subunit alpha 4; integrin, alpha 4; integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor); Itga4; Itga4B; ITGA-IV; LPAM subunit alpha; lymphocyte Peyer patch adhesion molecules subunit alpha; very late activation protein 4 receptor, alpha 4 subunit; VLA-4 receptor, alpha 4 subunit; VLA-4 subunit alpha | |
Mouse | |
Protein A | |
RUO | |
3676 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |