Learn More
ITGA3 Monoclonal Antibody (17C6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA181372
Code : LD
Additional Details : Weight : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD49c antibody, clone 17C6 recognizes CD49c, a approximately 150 kDa cell surface antigen which is also known as the alpha-3 integrin and as VLA-3.
ITGA3 Monoclonal antibody specifically detects ITGA3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen).Specifications
ITGA3 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P26006 | |
ITGA3 | |
CD49c transfected cell line. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen) | |
17C6 | |
Unconjugated | |
ITGA3 | |
AA407068; alpha 3 subunit of VLA-3 receptor; Alpha3 integrin; antigen CD49C; antigen identified by monoclonal antibody J143; CD49 antigen-like family member C; CD49c; CD49c antigen; FLJ34631; FLJ34704; FRP-2; Galactoprotein B3; GAPB3; GAP-B3; ILNEB; integrin alpha 3; Integrin alpha3; Integrin alpha-3; Integrin alpha-3 heavy chain; Integrin alpha-3 light chain; integrin subunit alpha 3; integrin, alpha 3; integrin, alpha 3 (antigen CD49C, alpha 3 subunit of VLA-3 receptor); ITGA3; MSK18; testicular tissue protein Li 96; testicular tissue protein Li 98; VCA-2; very late activation protein 3 receptor, alpha-3 subunit; VL3A; VLA-3 subunit alpha; VLA3a | |
Mouse | |
Protein A | |
RUO | |
3675 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |