missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD48 Monoclonal Antibody (OX-45), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517527
Code : NEW
Additional Details : Weight : 0.01000kg
Description
CD48 Monoclonal antibody specifically detects CD48 in Rat samples. It is validated for Flow Cytometry.Specifications
CD48 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P10252 | |
Cd48 | |
rat T cell blasts (stimulated purified T helper cells with allogeneic irradiated rat spleen cells) | |
500 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry | |
OX-45 | |
FITC | |
Cd48 | |
AI449234; AW610730; B-cell activation marker; BCM1; Bcm-1; BCM1 surface antigen; BLAST; BLAST1; BLAST-1; B-lymphocyte activation marker BLAST-1; Cd48; CD48 antigen; CD48 antigen (B cell membrane protein); CD48 antigen (B-cell membrane protein); Cd48 molecule; CD48d; hCD48; HM48-1; leukocyte antigen MEM-102; mCD48; MEM-102; MRC OX-45 surface antigen; sCD 48; sCD48; Sgp-60; Signaling lymphocytic activation molecule 2; Signaling lymphocytic activation molecule 2 (SLAMF2); SLAM family member 2; SLAMF2; soluble CD 48; soluble CD48; TCT.1 | |
Mouse | |
Protein G | |
RUO | |
245962 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |