Learn More
CD46 Monoclonal Antibody (MEM-258), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA182140
Code : LD
Additional Details : Weight : 0.01000kg
Description
This product contains sodium azide; removal by dialysis is recommended prior to use in functional assays. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD46 antibody, clone MEM-258 recognizes the human CD46 cell surface antigen, a broadly expressed glycoprotein also known as membrane co-factor protein (MCP).
CD46 Monoclonal antibody specifically detects CD46 in Human samples. It is validated for Flow Cytometry, Functional Assay, Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
CD46 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P15529 | |
CD46 | |
HPB-ALL cell line. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Functional Assay, Immunocytochemistry, Immunoprecipitation, Western Blot | |
MEM-258 | |
Unconjugated | |
CD46 | |
AHUS2; antigen identified by monoclonal antibody TRA-2-10; brain microvascular membrane cofactor; brain microvascular membrane cofactor protein CD46; Cd46; CD46 antigen, complement regulatory protein; CD46 complement regulatory protein; CD46 molecule; CD46 molecule, complement regulatory protein; CD46 splice variant; complement membrane cofactor protein; complement regulatory protein; Mcp; MCP; complement activation regulator; measles virus receptor; membrane cofactor protein; membrane cofactor protein (CD46, trophoblast-lymphocyte cross-reactive antigen); membrane cofactor protein variant 10; membrane cofactor protein variant 7; membrane cofactor protein variant 8; membrane cofactor protein variant 9; MGC26544; MIC10; soluble membrane cofactor protein variant 11; soluble membrane cofactor protein variant 12; soluble membrane cofactor protein variant 13; TLX; TRA2.10; trophoblast leucocyte common antigen; trophoblast leukocyte common antigen; trophoblast-lymphocyte cross-reactive antig | |
Mouse | |
Protein A | |
RUO | |
4179 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |