missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD46 Monoclonal Antibody (MEM-258), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119520
Code : LD
Additional Details : Weight : 0.01000kg
Description
CD46 Monoclonal antibody specifically detects CD46 in Bovine, Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Western Blot.Specifications
CD46 | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P15529, Q6VE48 | |
CD46 | |
T cell line HPB-ALL. | |
100 μg | |
Primary | |
Bovine, Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunoprecipitation, Western Blot | |
MEM-258 | |
Biotin | |
CD46 | |
AHUS2; antigen identified by monoclonal antibody TRA-2-10; brain microvascular membrane cofactor; brain microvascular membrane cofactor protein CD46; Cd46; CD46 antigen, complement regulatory protein; CD46 complement regulatory protein; CD46 molecule; CD46 molecule, complement regulatory protein; CD46 splice variant; complement membrane cofactor protein; complement regulatory protein; Mcp; MCP; complement activation regulator; measles virus receptor; membrane cofactor protein; membrane cofactor protein (CD46, trophoblast-lymphocyte cross-reactive antigen); membrane cofactor protein variant 10; membrane cofactor protein variant 7; membrane cofactor protein variant 8; membrane cofactor protein variant 9; MGC26544; MIC10; soluble membrane cofactor protein variant 11; soluble membrane cofactor protein variant 12; soluble membrane cofactor protein variant 13; TLX; TRA2.10; trophoblast leucocyte common antigen; trophoblast leukocyte common antigen; trophoblast-lymphocyte cross-reactive antig | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
280851, 4179 | |
4° C, do not freeze | |
Liquid |