missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Monoclonal Antibody (MEM-28), Alexa Fluor™ 488, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA518141
257.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This antibody reacts with all alternative forms of human CD45 antigen, and will not cross-react with equine.
CD45 Monoclonal antibody specifically detects CD45 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Specifications
CD45 | |
Monoclonal | |
Alexa Fluor 488 | |
PTPRC | |
B220; cd45; CD45 antigen; CD45 antigen isoform 1 precursor; CD45 antigen isoform 2 precursor; CD45 antigen isoform 3 precursor; CD45 antigen isoform 4 precursor; CD45 antigen isoform 5 precursor; CD45 antigen isoform 6 precursor; CD45R; GP180; Lca; L-CA; leucocyte common antigen; leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; leukocyte common antigen, CD45; loc; LOW QUALITY PROTEIN: receptor-type tyrosine-protein phosphatase C; LY5; Ly-5; lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase lambda; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; protein tyrosine phosphatase; alternatively spliced; Ptprc; Receptor-type tyrosine-protein phosphatase C; RT7; T200; T200 glycoprotein; T | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
5788 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry, Immunocytochemistry, Western Blot | |
MEM-28 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P08575 | |
PTPRC | |
Human thymocytes and T lymphocytes | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |