missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Monoclonal Antibody (OX-30), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517376
Code : NEW
Additional Details : Weight : 0.01000kg
Description
CD45 Monoclonal antibody specifically detects CD45 in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen).Specifications
CD45 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P04157 | |
PTPRC | |
Lymph Node glycoproteins and cells | |
500 μg | |
Primary | |
Rat | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Frozen) | |
OX-30 | |
Biotin | |
PTPRC | |
B220; cd45; CD45 antigen; CD45 antigen isoform 1 precursor; CD45 antigen isoform 2 precursor; CD45 antigen isoform 3 precursor; CD45 antigen isoform 4 precursor; CD45 antigen isoform 5 precursor; CD45 antigen isoform 6 precursor; CD45R; GP180; Lca; L-CA; leucocyte common antigen; leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; leukocyte common antigen, CD45; loc; LOW QUALITY PROTEIN: receptor-type tyrosine-protein phosphatase C; LY5; Ly-5; lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase lambda; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; protein tyrosine phosphatase; alternatively spliced; Ptprc; Receptor-type tyrosine-protein phosphatase C; RT7; T200; T200 glycoprotein; T | |
Mouse | |
Protein G | |
RUO | |
24699 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |