Learn More
CD45 Monoclonal Antibody (MEM-28), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119111
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody reacts with all alternative forms of human CD45 antigen, and will not cross-react with equine. Western Blot: non-reducing conditions, band around 200-250 kDa, 6% separating gel; Immunohistochemistry (Paraffin): No pre-treatment of tissue sections is needed; Immunocytochemistry: Paraformaldehyde fixation can be used.
CD45 Monoclonal antibody specifically detects CD45 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
CD45 | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P08575 | |
PTPRC | |
Human thymocytes and T lymphocytes. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
MEM-28 | |
Unconjugated | |
PTPRC | |
B220; cd45; CD45 antigen; CD45 antigen isoform 1 precursor; CD45 antigen isoform 2 precursor; CD45 antigen isoform 3 precursor; CD45 antigen isoform 4 precursor; CD45 antigen isoform 5 precursor; CD45 antigen isoform 6 precursor; CD45R; GP180; Lca; L-CA; leucocyte common antigen; leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; leukocyte common antigen, CD45; loc; LOW QUALITY PROTEIN: receptor-type tyrosine-protein phosphatase C; LY5; Ly-5; lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase lambda; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; protein tyrosine phosphatase; alternatively spliced; Ptprc; Receptor-type tyrosine-protein phosphatase C; RT7; T200; T200 glycoprotein; T | |
Mouse | |
Protein A | |
RUO | |
5788 | |
4° C, do not freeze | |
Liquid |